Please login to your online account to display your discounted pricing

UBP1112, Tocris

Group III mGlu antagonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material UBP1112
Assay Percent Range >99%
Molecular Formula C10H14NO5P
Formula Weight 259.2
Synonym α-Methyl-3-methyl-4-phosphonophenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

136910

 
r & d systems
RDS136910
10mg Each for $244.81

136950

 
r & d systems
RDS136950
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 259.2
Quantity 10mg
Chemical Name or Material UBP1112
CAS 339526-74-8
Assay Percent Range >99%
Molecular Formula C10H14NO5P
Synonym α-Methyl-3-methyl-4-phosphonophenylglycine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature