Please login to your online account to display your discounted pricing

UBP 302, Tocris

Selective, potent kainate antagonist; active enantiomer of UBP 296

$325.38 - $1,333.44

Chemical Identifiers

Chemical Name or Material UBP 302
Assay Percent Range >98%
Molecular Formula C15H15N3O6
Formula Weight 333.3
Synonym (S)-1-(2-Amino-2-carboxyethyl)-3-(2-carboxybenzyl)pyrimidine-2, 4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

207910

 
r & d systems
207910
10mg Each for $325.38

207950

 
r & d systems
207950
50mg Each for $1,333.44
Description & Specifications

Specifications

Formula Weight 333.3
Quantity 10mg
Assay Percent Range >98%
Synonym (S)-1-(2-Amino-2-carboxyethyl)-3-(2-carboxybenzyl)pyrimidine-2, 4-dione
Recommended Storage Store at Room Temperature
Chemical Name or Material UBP 302
Molecular Formula C15H15N3O6
CAS 745055-91-8
Safety and Handling

Store at Room Temperature