Please login to your online account to display your discounted pricing

UBP 296, Tocris

Selective, potent kainate antagonist; selective for GluR5-containing receptors

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material UBP 296
Assay Percent Range >98%
Molecular Formula C15H15N3O6
Formula Weight 333.3
Synonym (RS)-1-(2-Amino-2-carboxyethyl)-3-(2-carboxybenzyl)pyrimidine-2, 4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

207850

 
r & d systems
207850
50mg Each for $1,116.26

207810

 
r & d systems
207810
10mg Each for $265.15
Description & Specifications

Specifications

Formula Weight 333.3
Quantity 50mg
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Chemical Name or Material UBP 296
CAS 745055-86-1
Synonym (RS)-1-(2-Amino-2-carboxyethyl)-3-(2-carboxybenzyl)pyrimidine-2, 4-dione
Molecular Formula C15H15N3O6
Safety and Handling

Store at Room Temperature