Please login to your online account to display your discounted pricing

UBP 282, Tocris

AMPA/kainate antagonist; distinguishes between motoneuron and dorsal root kainate receptors

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material UBP 282
Assay Percent Range >98%
Molecular Formula C15H15N3O6
Formula Weight 333.3
Synonym (αS)-α-Amino-3-[(4-carboxyphenyl)methyl]-3, 4-dihydro-2, 4-dioxo-1(2H)-pyrimidinepropanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

176550

 
r & d systems
176550
50mg Each for $976.15

176510

 
r & d systems
176510
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 333.3
Quantity 50mg
Chemical Name or Material UBP 282
Assay Percent Range >98%
Molecular Formula C15H15N3O6
CAS 544697-47-4
Synonym (αS)-α-Amino-3-[(4-carboxyphenyl)methyl]-3, 4-dihydro-2, 4-dioxo-1(2H)-pyrimidinepropanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature