missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ubiquitin Mouse, Clone: eBioP4D1 (P4D1), eBioscience™

Mouse Monoclonal Antibody

$104.28 - $223.08

Specifications

Antigen Ubiquitin
Clone eBioP4D1 (P4D1)
Host Species Murine
Applications Immunohistochemistry,Immunoprecipitation,Western Blot
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013455 Life Technologies
LS14607880
25μg Each for $104.28
5013456 Life Technologies
LS14607882
100μg Each for $223.08
Specifications

Description

The monoclonal antibody eBioP4D1 (p4D1) recognizes ubiquitin, a small 8Ka protein. Ubiquitin units are linked to proteins via a ubiquitination process, thereby signaling the protein for degradation by the 26S proteosome. The process of ubiquitination plays a role in differentiation, cell cycle progression, apoptosis and disease progression. For example, neuronal inclusions called Lewis bodies from Parkinson patients are rich in ubiquitin.
Description & Specifications

Specifications

Ubiquitin
Murine
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioP4D1 (P4D1)
Immunohistochemistry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL