Please login to your online account to display your discounted pricing

Ubiquitin Mouse, Biotin, Clone: eBioP4D1 (P4D1), eBioscience™

Mouse Monoclonal Antibody

$129.05 - $274.05

Specifications

Antigen Ubiquitin
Clone eBioP4D1 (P4D1)
Host Species Mouse
Applications Western Blotting
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012129

 
affymetrix
13607880
25μg Each for $129.05

5012130

 
affymetrix
13607882
100μg Each for $274.05
Description & Specifications

Specifications

Antigen Ubiquitin
Clone eBioP4D1 (P4D1)
Host Species Mouse
Applications Western Blotting
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody eBioP4D1 (p4D1) recognizes ubiquitin, a small 8Ka protein. Ubiquitin units are linked to proteins via a ubiquitination process, thereby signaling the protein for degradation by the 26S proteosome. The process of ubiquitination plays a role in differentiation, cell cycle progression, apoptosis and disease progression. For example, neuronal inclusions called Lewis bodies from Parkinson patients are rich in ubiquitin.