Please login to your online account to display your discounted pricing

Tocris™ UAMC 00039 dihydrochloride

Dipeptidyl peptidase II (DPP-II) inhibitor

$283.92 - $1,179.73

Specifications

Product Type UAMC 00039 dihydrochloride
Inhibitors UAMC 00039 dihydrochloride
Molecular Weight 382.76
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

495410R

 
tocris™
RDS495410
10mg Each for $283.92

495450R

 
tocris™
RDS495450
50mg Each for $1,179.73
Description & Specifications

Specifications

Product Type UAMC 00039 dihydrochloride
Inhibitors UAMC 00039 dihydrochloride
Molecular Weight 382.76
Purity >98

Potent inhibitor of dipeptidyl peptidase II (DPP-II) (IC50 = 0.48nM). Exhibits selectivity for DPP-II against DPP-9, DPP-8 and DPP-IV (IC50 values are 78.6, 142 and 165μM, respectively). Orally available.

  • Chemical Name: (2S)-2-Amino-4-[[(4-chlorophenyl)methyl]amino]-1-(1-piperidinyl)-1-butanone dihydrochloride
  • Formula: C16H24ClN3O.2HCl
  • CAS Number: 697797-51-6
  • Solubility: Soluble to 100mM in water and to 100mM in DMSO
  • Purity: >98