Please login to your online account to display your discounted pricing

U0126, Tocris


Potent, selective inhibitor of MEK1 and 2

Quantity 25mg

Manufacturer: r & d systems  RDS114425

 View more versions of this product

Catalog No. 114425

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 380.48
Quantity 25mg
Chemical Name or Material U0126
Assay Percent Range >98%
Molecular Formula C18H16N6S2
CAS 109511-58-2
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis[2-aminophenylthio]butadiene
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material U0126
Assay Percent Range >98%
Molecular Formula C18H16N6S2
Formula Weight 380.48
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis[2-aminophenylthio]butadiene
Accessories

Desiccate at +4°C