Please login to your online account to display your discounted pricing

U0126, Tocris

Potent, selective inhibitor of MEK1 and 2

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material U0126
Assay Percent Range >98%
Molecular Formula C18H16N6S2
Formula Weight 380.48
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis[2-aminophenylthio]butadiene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

114425

 
r & d systems
RDS114425
25mg Each for $1,050.42

11445

  SDS
r & d systems
RDS11445
5mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 380.48
Quantity 25mg
CAS 109511-58-2
Chemical Name or Material U0126
Molecular Formula C18H16N6S2
Assay Percent Range >98%
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis[2-aminophenylthio]butadiene
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C