Please login to your online account to display your discounted pricing

U 99194 maleate, Tocris


Potent, selective D3 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS135710

 View more versions of this product

Catalog No. 135710

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 393.48
Quantity 10mg
Chemical Name or Material U 99194 maleate
Molecular Formula C17H27NO2·C4H4O4
CAS 234757-41-6
Synonym 2,3-Dihydro-5,6-dimethoxy-N, N-dipropyl-1H-inden-2-amine maleate
Recommended Storage Store at Room Temperature
Assay Percent Range >98%

Specifications

Chemical Name or Material U 99194 maleate
Assay Percent Range >98%
Molecular Formula C17H27NO2·C4H4O4
Formula Weight 393.48
Synonym 2,3-Dihydro-5,6-dimethoxy-N, N-dipropyl-1H-inden-2-amine maleate
Accessories

Store at Room Temperature