Please login to your online account to display your discounted pricing

U 99194 maleate, Tocris

Potent, selective D3 antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material U 99194 maleate
Assay Percent Range >98%
Molecular Formula C17H27NO2·C4H4O4
Formula Weight 393.48
Synonym 2,3-Dihydro-5,6-dimethoxy-N, N-dipropyl-1H-inden-2-amine maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

135710

 
r & d systems
RDS135710
10mg Each for $177.37

135750

 
r & d systems
RDS135750
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 393.48
Quantity 10mg
Chemical Name or Material U 99194 maleate
Molecular Formula C17H27NO2·C4H4O4
Recommended Storage Store at Room Temperature
CAS 234757-41-6
Synonym 2,3-Dihydro-5,6-dimethoxy-N, N-dipropyl-1H-inden-2-amine maleate
Assay Percent Range >98%
Safety and Handling

Store at Room Temperature