Please login to your online account to display your discounted pricing

U 93631, Tocris

GABAA receptor antagonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material U 93631
Assay Percent Range >99%
Molecular Formula C17H21N3O2
Formula Weight 299.37
Synonym 4,5-Dihydro-4,4-dimethyl-imidazo[1,5-a]quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

274510

 
r & d systems
274510
10mg Each for $225.77

274550

 
r & d systems
274550
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 299.37
Quantity 10mg
Chemical Name or Material U 93631
Assay Percent Range >99%
CAS 152273-12-6
Synonym 4,5-Dihydro-4,4-dimethyl-imidazo[1,5-a]quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester
Recommended Storage Store at Room Temperature
Molecular Formula C17H21N3O2
Safety and Handling

Store at Room Temperature