Please login to your online account to display your discounted pricing

Alfa Aesar™ U-83836E, 98%


Manufacturer: alfa aesar™  J67472MA

 View more versions of this product

Catalog No. AAJ67472MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Assay 98%
CAS 122003-11-6
Molecular Formula C30H44N6O2.2HCl
Formula Weight 593.63
Quantity 10mg
Assay 98%
Description White
Storage Requirements -30°C to -10°C
Format Solid
Type U-83836E, 98%