Please login to your online account to display your discounted pricing

U 73122, Tocris


Phospholipase C inhibitor

Quantity 10mg

Manufacturer: r & d systems  126810

 View more versions of this product

Catalog No. 126810

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 464.65
Quantity 10mg
Chemical Name or Material U 73122
Assay Percent Range >98%
Molecular Formula C29H40N2O3
CAS 112648-68-7
Synonym 1-[6-[[(17β)-3-Methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]-1H-pyrrole-2,5-dione
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material U 73122
Assay Percent Range >98%
Molecular Formula C29H40N2O3
Formula Weight 464.65
Synonym 1-[6-[[(17β)-3-Methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]-1H-pyrrole-2,5-dione
Accessories

Store at Room Temperature