Please login to your online account to display your discounted pricing

U 73122, Tocris

Phospholipase C inhibitor

$175.83 - $734.99

Chemical Identifiers

Chemical Name or Material U 73122
Assay Percent Range >98%
Molecular Formula C29H40N2O3
Formula Weight 464.65
Synonym 1-[6-[[(17β)-3-Methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]-1H-pyrrole-2,5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

126810

 
r & d systems
126810
10mg Each for $175.83

126850

 
r & d systems
126850
50mg Each for $734.99
Description & Specifications

Specifications

Chemical Name or Material U 73122
Quantity 10mg
CAS 112648-68-7
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C29H40N2O3
Formula Weight 464.65
Synonym 1-[6-[[(17β)-3-Methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]-1H-pyrrole-2,5-dione
Safety and Handling

Store at Room Temperature