Please login to your online account to display your discounted pricing

U-54494A hydrochloride, Tocris


κ agonist

Quantity 50mg

Manufacturer: r & d systems  049850

 View more versions of this product

Catalog No. 049850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 391.77
Chemical Name or Material U-54494A hydrochloride
Assay Percent Range >98%
Molecular Formula C18H24Cl2N2O·HCl
CAS 112465-94-8
Synonym cis-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzamide hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material U-54494A hydrochloride
Assay Percent Range >98%
Molecular Formula C18H24Cl2N2O·HCl
Formula Weight 391.77
Synonym cis-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzamide hydrochloride
Accessories

Store at Room Temperature