missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-U-50488 hydrochloride, Tocris

Standard selective κ agonist

$181.19 - $762.58

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C19H26Cl2N2O·HCl
Formula Weight 405.79
Synonym trans-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride
Chemical Name or Material (±)-U-50488 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
047150 R & D Systems
047150
50mg Each for $762.58
047110 R & D Systems
047110
10mg Each for $181.19
049525 R & D Systems
049525
25mg Each for $181.19
Specifications Description & Specifications

Chemical Identifiers

>99%
405.79
(±)-U-50488 hydrochloride
C19H26Cl2N2O·HCl
trans-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride

Specifications

50mg
>99%
405.79
(±)-U-50488 hydrochloride
67197-96-0
C19H26Cl2N2O·HCl
trans-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride

Safety and Handling

Recommended Storage : Desiccate at Room Temperature