Please login to your online account to display your discounted pricing

(-)-U-50488 hydrochloride, Tocris

Standard selective κ agonist; More active enantiomer of (±)-U-50488

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material (-)-U-50488 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H26Cl2N2O·HCl
Formula Weight 405.79
Synonym trans-(-)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

049650

 
r & d systems
049650
50mg Each for $812.12

049610

 
r & d systems
049610
10mg Each for $192.67
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 405.79
Chemical Name or Material (-)-U-50488 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H26Cl2N2O·HCl
CAS 114528-79-9
Synonym trans-(-)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature