Please login to your online account to display your discounted pricing

(±)-U-50488 hydrochloride, Tocris


Standard selective κ agonist

Quantity 50mg

Manufacturer: r & d systems  047150

 View more versions of this product

Catalog No. 047150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 405.79
Chemical Name or Material (±)-U-50488 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H26Cl2N2O·HCl
CAS 67197-96-0
Synonym trans-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material (±)-U-50488 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H26Cl2N2O·HCl
Formula Weight 405.79
Synonym trans-(±)-3, 4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide hydrochloride
Accessories

Desiccate at Room Temperature