Please login to your online account to display your discounted pricing

Alfa Aesar™ U-18666A, 95%


Manufacturer: alfa aesar™  J67389LB0

 View more versions of this product

Catalog No. AAJ67389LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

CAS 3039-71-2
Formula Weight 424.06
Description White to off-white
Storage Requirements 2°C to 8°C
Type U-18666A, 95%
Assay 95%
Solubility Soluble in water at 10mg/ml
Quantity 5mg
Synonym 3β-[2-(Diethylamino)ethoxy]androstenone hydrochloride
Melting Point 195-196.5 
Format Solid
Molecular Formula C25H41NO2.HCl