Please login to your online account to display your discounted pricing

Alfa Aesar™ U-0521, 98%


Manufacturer: alfa aesar™  J67355FPL

 View more versions of this product

Catalog No. AAJ67355FPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Assay 98%
CAS 5466-89-7
Molecular Formula C10H12O3
Formula Weight 180.20
Synonym 3',4'-Dihydroxy-2-methylpropiophenone;
Quantity 20mg
Melting Point 79-81°
Assay 98%
Description White
Format Solid
Type U-0521, 98%