missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals U-0521, 98%
$207.99 - $207.99
Chemical Identifiers
CAS | 5466-89-7 |
---|---|
Molecular Formula | C10H12O3 |
Synonym | 3',4'-Dihydroxy-2-methylpropiophenone; |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67355FPL
|
Thermo Scientific Chemicals
J67355FPL |
20 mg |
Each for $207.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5466-89-7 | |
C10H12O3 | |
3',4'-Dihydroxy-2-methylpropiophenone; |
Specifications
20 mg | |
5466-89-7 | |
180.20 | |
79°C to 81°C | |
Solid |
White | |
C10H12O3 | |
3',4'-Dihydroxy-2-methylpropiophenone; | |
U-0521, 98% | |
98% |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only