missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tyvek™ Lab Coats
Disposable, yet strong enough to withstand several wearings.
Manufacturer: Fisher Science Education 01212WHLGE
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Lightweight and tear-resistant Tyvek fabric
- Knee-length with two pockets, a five-snap front and full sleeves
- Inherent barrier protection against dry particulate hazards
- Chemically and biologically inert
- Provide limited splash protection
- White
Specifications
Yes | |
Mandarin | |
White | |
Tyvek™ fabric | |
Large | |
83.82cm | |
38.25 to 41.75 in. |
Knee length | |
Snaps | |
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up | |
2 | |
33 in. | |
Serged |