DuPont™ Tyvek 400 Lab Coats, TY210
Disposable, yet strong enough to withstand several wearings.
$198.26 - $241.43
Specifications
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
View More Specs
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
01-361-11A
View Documents Promotion Details
DuPont™
TY210SWHSM003000
Small
Case of 30 Each for $203.61
01-361-11B
View Documents Promotion Details
DuPont™
TY210SWHMD003000
Medium
Case of 30 Each for $205.80
01-361-11C
View Documents Promotion Details
DuPont™
TY210SWHLG003000
Large
Case of 30 Each for $204.01
19087223
View Documents Promotion Details
DuPont™
TY210SWHXL003000
X-Large
Case of 30 Each for $198.26
19087224
View Documents Promotion Details
DuPont™
TY210SWH2X003000
2X-Large
Case of 30 Each for $210.08
19-087-225
View Documents Promotion Details
DuPont™
TY210SWH3X003000
3X-Large
Case of 30 Each for $241.43
Description
Description
Disposable, yet strong enough to withstand several wearings
  • Lightweight and tear-resistant Tyvek fabric
  • Knee length with five-snap front and full sleeves
  • Chemically and biologically inert
  • Provide limited splash protection
  • White
Specifications
Specifications
Tyvek
Snaps
Single-use
Lay down
Lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up
White
Open
Knee-length
Serged
Bulk
SDS
missing translation for 'documents'
missing translation for 'documents'