Please login to your online account to display your discounted pricing

Alfa Aesar™ Tyrphostin RG 14620, 98%

Specifications

Description White
Format Solid
Assay 98%
CAS 136831-49-7
Molecular Formula C14H8Cl2N2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67185LB0

 
alfa aesar™
J67185LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67185MA

 
alfa aesar™
J67185MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Description White
Format Solid
Assay 98%
CAS 136831-49-7
Molecular Formula C14H8Cl2N2
Formula Weight 275.14g/mol
Melting Point 150° to 151°C
Sensitivity Light Sensitive
Synonym 3-(3,5-Dichlorophenyl)-2-(3-pyridyl)acrylonitrile; RG-14620