missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin B46, 98+%, Thermo Scientific Chemicals
Inhibitor of EGF receptor kinase activity
$307.16 - $307.16
Chemical Identifiers
| CAS | 133550-34-2 |
|---|---|
| Molecular Formula | C19H18N2O3 |
| Molecular Weight (g/mol) | 322.364 |
| MDL Number | MFCD00209865 |
| InChI Key | GSQOBTOAOGXIFL-WJDWOHSUSA-N |
| Synonym | tyrphostin b46, z-2-cyano-3-3,4-dihydroxyphenyl-n-3-phenylpropyl prop-2-enamide, n-3 inverted exclamation marka-phenylpropyl-3,4-dihydroxybenzylidenecyanoacetamide |
| PubChem CID | 5353390 |
| IUPAC Name | (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide |
| SMILES | C1=CC=C(C=C1)CCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61704MA
|
Thermo Scientific Chemicals
J61704MA |
10 mg |
Each for $307.16
|
|
|||||
Description
Inhibitor of EGF receptor kinase activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 133550-34-2 | |
| 322.364 | |
| GSQOBTOAOGXIFL-WJDWOHSUSA-N | |
| 5353390 | |
| C1=CC=C(C=C1)CCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N |
| C19H18N2O3 | |
| MFCD00209865 | |
| tyrphostin b46, z-2-cyano-3-3,4-dihydroxyphenyl-n-3-phenylpropyl prop-2-enamide, n-3 inverted exclamation marka-phenylpropyl-3,4-dihydroxybenzylidenecyanoacetamide | |
| (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide |
Specifications
| 133550-34-2 | |
| C19H18N2O3 | |
| 10 mg | |
| GSQOBTOAOGXIFL-WJDWOHSUSA-N | |
| (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide | |
| 5353390 | |
| ≥98% | |
| Tyrphostin B46 |
| Yellow | |
| MFCD00209865 | |
| tyrphostin b46, z-2-cyano-3-3,4-dihydroxyphenyl-n-3-phenylpropyl prop-2-enamide, n-3 inverted exclamation marka-phenylpropyl-3,4-dihydroxybenzylidenecyanoacetamide | |
| C1=CC=C(C=C1)CCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N | |
| 322.364 | |
| 322.36 | |
| Powder |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only