Please login to your online account to display your discounted pricing

Tyrphostin B44, (-) enantiomer, Tocris


EGFR-kinase inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS057810

 View more versions of this product

Catalog No. 057810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 308.34
CAS 133550-32-0
Chemical Name or Material Tyrphostin B44, (-) enantiomer
Assay Percent Range >98%
Molecular Formula C18H16N2O3
Synonym (R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Tyrphostin B44, (-) enantiomer
Assay Percent Range >98%
Molecular Formula C18H16N2O3
Formula Weight 308.34
Synonym (R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Accessories

Desiccate at +4°C