Please login to your online account to display your discounted pricing

Tyrphostin B44, (-) enantiomer, Tocris

EGFR-kinase inhibitor

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material Tyrphostin B44, (-) enantiomer
Assay Percent Range >98%
Molecular Formula C18H16N2O3
Formula Weight 308.34
Synonym (R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

057810

 
r & d systems
RDS057810
10mg Each for $129.05

057850

 
r & d systems
RDS057850
50mg Each for $546.19
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 308.34
CAS 133550-32-0
Chemical Name or Material Tyrphostin B44, (-) enantiomer
Assay Percent Range >98%
Molecular Formula C18H16N2O3
Synonym (R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C