Please login to your online account to display your discounted pricing

Tyrphostin B44, (+) enantiomer, Tocris

EGFR-kinase inhibitor

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material Tyrphostin B44, (+) enantiomer
Assay Percent Range >98%
Molecular Formula C18H16N2O3
Formula Weight 308.34
Synonym (S)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

057950

 
r & d systems
RDS057950
50mg Each for $618.87

057910

 
r & d systems
RDS057910
10mg Each for $146.74
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 308.34
Assay Percent Range >98%
Chemical Name or Material Tyrphostin B44, (+) enantiomer
Molecular Formula C18H16N2O3
CAS 133550-37-5
Synonym (S)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C