Tyrphostin B44, (+) enantiomer, Tocris
EGFR-kinase inhibitor
Manufacturer:  R & D Systems RDS057910
 View more versions of this product
Catalog No. 057910
Specifications Description & Specifications
Specifications
Tyrphostin B44, (+) enantiomer
>98%
308.34
10mg
133550-37-5
C18H16N2O3
(S)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Chemical Identifiers
>98%
308.34
Tyrphostin B44, (+) enantiomer
C18H16N2O3
(S)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Safety and Handling
Recommended Storage : Desiccate at +4°C