missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tyrphostin B42, 99+%

A protein kinase inhibitor

$249.82 - $462.64

Chemical Identifiers

CAS 134036-52-5
Molecular Formula C17H14N2O3
Molecular Weight (g/mol) 294.31
MDL Number MFCD00236452
InChI Key TUCIOBMMDDOEMM-ZSOIEALJSA-N
Synonym 2-cyano-3-3,4-dihydroxyphenyl-n-phenylmethyl-2-propenamide, 2-cyano-3-3,4-dihydroxyphenyl-n-benzyl-2-propenamide, z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, 2z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide
PubChem CID 5353441
IUPAC Name (Z)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
SMILES C1=CC=C(C=C1)CNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61715MB View Documents Alfa Aesar™
J61715MB
25mg Each for $249.82
 
AAJ61715MC View Documents Alfa Aesar™
J61715MC
100mg Each for $462.64
 
Description

Description

Applications
Tyrphostin B42 is used in acute lymphoblastic leukemia studies the compound has been reported to inhibit JAK2 from B cell precursors.

Solubility
Soluble in DMSO or ethanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

134036-52-5
294.31
TUCIOBMMDDOEMM-ZSOIEALJSA-N
5353441
C1=CC=C(C=C1)CNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
C17H14N2O3
MFCD00236452
2-cyano-3-3,4-dihydroxyphenyl-n-phenylmethyl-2-propenamide, 2-cyano-3-3,4-dihydroxyphenyl-n-benzyl-2-propenamide, z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, 2z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide
(Z)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide

Specifications

Green-Yellow
25mg
C17H14N2O3
2-cyano-3-3,4-dihydroxyphenyl-n-phenylmethyl-2-propenamide, 2-cyano-3-3,4-dihydroxyphenyl-n-benzyl-2-propenamide, z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, 2z-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide
TUCIOBMMDDOEMM-ZSOIEALJSA-N
(Z)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
5353441
Powder
Tyrphostin B42
216°C to 220°C
134036-52-5
MFCD00236452
Soluble in DMSO or ethanol
C1=CC=C(C=C1)CNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
294.31
294.31
≥99%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C