Please login to your online account to display your discounted pricing

Alfa Aesar™ Tyrphostin AG-213, 97%


Manufacturer: alfa aesar™  J67345LB0

 View more versions of this product

Catalog No. AAJ67345LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Formula Weight 220.25
Chemical Name or Material Tyrphostin AG-213
Assay 97%
CAS 122520-86-9
Molecular Formula C10H8N2O2S
Quantity 5mg
Assay 97%
MDL Number MFCD00133902
Description Orange
Synonym RG-50864; Tyrphostin 47
Physical Form Solid
Color Orange
Formula Weight 220.25g/mol
Quantity 5mg
Format Solid