Please login to your online account to display your discounted pricing

Alfa Aesar™ Tyrphostin AG-183, 98%


Manufacturer: alfa aesar™  J67135MA

 View more versions of this product

Catalog No. AAJ67135MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

CAS 122520-90-5
Type Tyrphostin AG-183, 98%
Formula Weight 268.23
Assay 98%
Format Solid
Synonym 2-Amino-1,1,3-tricyano-4-(3',4',5'-trihydroxyphenyl)butadiene; Tyrphostin 51
Molecular Formula C13H8N4O3
Quantity 10mg
Storage Requirements -30°C to -10°C
Description Brown