Please login to your online account to display your discounted pricing

Alfa Aesar™ Tyrphostin AG-1296, 98%


Quantity 5mg

Manufacturer: alfa aesar™  J67039LB0

 View more versions of this product

Catalog No. AAJ67039LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 266.3
Assay 98%
MDL Number MFCD00270913
CAS 146535-11-7
Physical Form Solid
Synonym 6,7-Dimethoxy-3-phenylquinoxaline
Color Yellow
Quantity 5mg
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C
Chemical Name or Material Tyrphostin AG-1296

Specifications

Chemical Name or Material Tyrphostin AG-1296
CAS 146535-11-7
Formula Weight 266.3
MDL Number MFCD00270913
Synonym 6,7-Dimethoxy-3-phenylquinoxaline
Accessories

-30° to -10°C