Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.

$268.31 - $1,353.39

Specifications

Antigen Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1
Clone TA99
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Applications Immunohistochemistry (Frozen)
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1634P

 
thermo scientific™ lab vision™
MS1634P
200μg/mL 1mL Each for $1,269.04

MS1634P0

 
thermo scientific™ lab vision™
MS1634P0
200μg/mL 100µL Each for $268.31

MS1634P1

 
thermo scientific™ lab vision™
MS1634P1
200μg/mL 500µL Each for $759.78

MS1634P1ABX

 
thermo scientific™ lab vision™
MS1634P1ABX
1mg/mL 100µL Each for $843.69

MS1634PABX

 
thermo scientific™ lab vision™
MS1634PABX
1mg/mL 200µL Each for $1,353.39
Description & Specifications

Specifications

Antigen Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1
Clone TA99
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Enzyme
Type Primary
Immunogen SK-MEL-23 Melanoma cell line
Isotype IgG2a

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.

Tyrosinase related Protein 75 (TRP75/gp75) is a pigmentation associated protein expressed by melanoma cells and normal melanocytes. This antigen appears to be expressed during stage III and IV of melanosomes.

Host Species: Mouse

Clone: TA99

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: SK-MEL-23 Melanoma cell line

Molecular Weight: 75kDa

Positive Control: SK-MEL-23, A375 cells; Melanoma, Skin

Cellular Localization: Cytoplasmic Ab-1 is specific for melanomas and does not react with carcinomas and sarcomas.

Recommended for:

  • Western Blotting
  • Immunohistochemistry (Frozen)

Note:

USA: RUO; Int'l: RUO