Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1634P1ABX

 View more versions of this product

Catalog No. MS1634P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Immunogen SK-MEL-23 Melanoma cell line
Regulatory Status RUO
Antigen Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Clone TA99
Host Species Mouse
Isotype IgG2a
Format Concentrated
Quantity 100µL
Conjugate Unlabeled
Classification Monoclonal
Research Discipline Enzyme
Type Primary
Concentration 1mg/mL

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.

Tyrosinase related Protein 75 (TRP75/gp75) is a pigmentation associated protein expressed by melanoma cells and normal melanocytes. This antigen appears to be expressed during stage III and IV of melanosomes.

Host Species: Mouse

Clone: TA99

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: SK-MEL-23 Melanoma cell line

Molecular Weight: 75kDa

Positive Control: SK-MEL-23, A375 cells; Melanoma, Skin

Cellular Localization: Cytoplasmic Ab-1 is specific for melanomas and does not react with carcinomas and sarcomas.

Recommended for:

  • Western Blotting
  • Immunohistochemistry (Frozen)

Note:

USA: RUO; Int'l: RUO