missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1634PABX

 View more versions of this product

Catalog No. MS1634PABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1, Mouse Monoclonal Antibody.

Tyrosinase related Protein 75 (TRP75/gp75) is a pigmentation associated protein expressed by melanoma cells and normal melanocytes. This antigen appears to be expressed during stage III and IV of melanosomes.

Host Species: Mouse

Clone: TA99

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: SK-MEL-23 Melanoma cell line

Molecular Weight: 75kDa

Positive Control: SK-MEL-23, A375 cells; Melanoma, Skin

Cellular Localization: Cytoplasmic Ab-1 is specific for melanomas and does not react with carcinomas and sarcomas.

Recommended for:

  • Western Blotting
  • Immunohistochemistry (Frozen)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Tyrosinase Related Protein 75 (TRP75/gp75) Ab-1
Monoclonal
1mg/mL
Concentrated
SK-MEL-23 Melanoma cell line
200μL
Enzyme
Monoclonal
Immunohistochemistry (Frozen),Western Blotting
TA99
Unlabeled
Murine
IgG2a
RUO
Primary
Human