missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Human TYRO3 (RSE), GST Tag Recombinant Protein
Human TYRO3 (RSE), GST Tag Recombinant Protein
Supplier: Thermo Fisher Scientific PV3828
Description
TYRO3 is a receptor tyrosine kinase that has been shown to be involved in signal transduction and cell adhesion.Specifications
NP_006284 | |
7301 | |
10 μg | |
AXL Kinase Family | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
TYRO3 | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
76.6 kDa | |
TYRO3 (RSE), DKT, BRT, TIF | |
aa 451-890 | |
Recombinant | |
Storage Buffer: 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
TYRO3 protein tyrosine kinase | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |