missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[Tyr8, Nle11] Substance P, Alfa Aesar™

For cellular and molecular biology applications

Specifications

Product Type [Tyr8, Nle11] Substance P
Molecular Weight 1345.61
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Lys-Pro-Gln-Gln-Phe-Tyr-Gly-Leu-Nle-NH2
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66679LB0
View Documents
Thermo Fisher Scientific
J66679LB0
5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66679MA
View Documents
Thermo Fisher Scientific
J66679MA
10mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

[Tyr8, Nle11] Substance P
-30° to -10°C
Solid
HPLC
1345.61
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Tyr-Gly-Leu-Nle-NH2
RUO

For Research Use Only