Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Neurokinin B

For cellular and molecular biology applications

Specifications

Product Type [Tyr0] Neurokinin B
Molecular Weight 1373.61
Storage Requirements -30° to -10°C
Sequence Tyr-Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66585LB0

 
alfa aesar™
J66585LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr0] Neurokinin B
Molecular Weight 1373.61
Storage Requirements -30° to -10°C
Sequence Tyr-Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC