Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Neurokinin A

For cellular and molecular biology applications

Specifications

Product Type [Tyr0] Neurokinin A
Molecular Weight 1296.51
Storage Requirements -30° to -10°C
Format Solid
Sequence Tyr-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66948EXD

 
alfa aesar™
J66948EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66948MCR

 
alfa aesar™
J66948MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr0] Neurokinin A
Molecular Weight 1296.51
Storage Requirements -30° to -10°C
Format Solid
Sequence Tyr-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
Regulatory Status RUO
Purification Method HPLC