Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Neurokinin A


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66948EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66948EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Product Type [Tyr0] Neurokinin A
Sequence Tyr-His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
Purification Method HPLC
Molecular Weight 1296.51
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO