Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Fibrinopeptide A, Human

For cellular and molecular biology applications

Specifications

Product Type [Tyr0] Fibrinopeptide A, human
Molecular Weight 1699.73
Storage Requirements -30° to -10°C
Sequence Tyr-Ala-Asp-Ser-Gly-Glu-Gly-Asp-Phe-Leu-Ala-Glu-Gly-Gly-Gly-Val-Arg
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66132LB0

 
alfa aesar™
J66132LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66132MA

 
alfa aesar™
J66132MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr0] Fibrinopeptide A, human
Molecular Weight 1699.73
Storage Requirements -30° to -10°C
Sequence Tyr-Ala-Asp-Ser-Gly-Glu-Gly-Asp-Phe-Leu-Ala-Glu-Gly-Gly-Gly-Val-Arg
Format Solid
Regulatory Status RUO
Purification Method HPLC