Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_6;, D-Phe&superscript_7;, D-His&superscript_9;] Substance P (6-11)

For cellular and molecular biology applications

Specifications

Product Type [Tyr6, D-Phe7, D-His9] Substance P (6-11)
Molecular Weight 856.05
Storage Requirements -30° to -10°C
Format Solid
Common Name Sendide
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66739LB0

 
alfa aesar™
J66739LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66739MA

 
alfa aesar™
J66739MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr6, D-Phe7, D-His9] Substance P (6-11)
Molecular Weight 856.05
Storage Requirements -30° to -10°C
Format Solid
Common Name Sendide
Regulatory Status RUO
Purification Method HPLC