missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[Tyr4, D-Phe12] Bombesin, Alfa Aesar™

For cellular and molecular biology applications

$96.85 - $315.19

Specifications

Product Type [Tyr4, D-Phe12] Bombesin
Molecular Weight 1679.89
Storage Requirements -30° to -10°C
Sequence Glp-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-D_Phe-Leu-Met-NH2
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66313MCR
View Documents
Thermo Fisher Scientific
J66313MCR
1mg Each for $96.85
 
AAJ66313LB0
View Documents
Thermo Fisher Scientific
J66313LB0
5mg Each for $315.19
 
Specifications

Specifications

[Tyr4, D-Phe12] Bombesin
-30° to -10°C
Solid
RUO
1679.89
Glp-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-D_Phe-Leu-Met-NH2
Bombesin Receptor Antagonist
HPLC

For Research Use Only