Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Calcitonin Gene Related Peptide, Rat

For cellular and molecular biology applications

Specifications

Product Type [Tyr0] Calcitonin Gene Related Peptide, rat
Molecular Weight 3969.41
Storage Requirements -30° to -10°C
Format Solid
Sequence [Tyr0] CGRP, rat
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66788EXD

 
alfa aesar™
J66788EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66788MCR

 
alfa aesar™
J66788MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr0] Calcitonin Gene Related Peptide, rat
Molecular Weight 3969.41
Storage Requirements -30° to -10°C
Format Solid
Sequence [Tyr0] CGRP, rat
Regulatory Status RUO
Purification Method HPLC