Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Brain Natriuretic Peptide (1-32), Human

For cellular and molecular biology applications

Specifications

Product Type [Tyr0] Brain Natriuretic Peptide (1-32), human
Cross Reactivity Human
Molecular Weight 3627.20
Storage Requirements -30° to -10°C
Sequence [Tyr0] BNP (1-32), human
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66123EXD

 
alfa aesar™
J66123EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66123MCR

 
alfa aesar™
J66123MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr0] Brain Natriuretic Peptide (1-32), human
Cross Reactivity Human
Molecular Weight 3627.20
Storage Requirements -30° to -10°C
Sequence [Tyr0] BNP (1-32), human
Format Solid
Regulatory Status RUO
Purification Method HPLC