Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_4;] Bombesin

For cellular and molecular biology applications

$86.20 - $203.19

Specifications

Product Type [Tyr4] Bombesin
Molecular Weight 1669.86
Storage Requirements -30° to -10°C
Sequence Glp-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66705LB0

 
alfa aesar™
J66705LB0
5mg Each for $203.19

AAJ66705MCR

 
alfa aesar™
J66705MCR
1mg Each for $86.20
Description & Specifications

Specifications

Product Type [Tyr4] Bombesin
Molecular Weight 1669.86
Storage Requirements -30° to -10°C
Sequence Glp-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC