Please login to your online account to display your discounted pricing

Alfa Aesar™ [Tyr&superscript_0;] Atrial Natriuretic Peptide II, Rat

For cellular and molecular biology applications

Specifications

Product Type [Tyr0] Atrial Natriuretic Peptide II, rat
Cross Reactivity Rat
Molecular Weight 2549.79
Storage Requirements -30° to -10°C
Sequence [Tyr0] Atriopeptin II
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66693EXD

 
alfa aesar™
J66693EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66693MCR

 
alfa aesar™
J66693MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Tyr0] Atrial Natriuretic Peptide II, rat
Cross Reactivity Rat
Molecular Weight 2549.79
Storage Requirements -30° to -10°C
Sequence [Tyr0] Atriopeptin II
Format Lyophilized powder
Common Name [Tyr126] ANP (126-149), rat
Regulatory Status RUO
Purification Method HPLC