Invitrogen™ TYK2 Recombinant Human Protein DFS Item
Tyrosine Kinase 2 (TYK2) is a member of the JAK kinase family
Manufacturer:  Invitrogen™ LSPV4791
 View more versions of this product
Catalog No. PV4791
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Tyrosine Kinase 2 (TYK2) plays both structural and catalytic roles in type I interferon (IFN) signaling.
Specifications
Specifications
NP_003322.2
Frozen
68.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
JAK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% glycerol
1mg
RUO - research use only
TYK2
Baculovirus
Human
GST Tag
Documents